ENA369383378 Enamine

methyl 1-aminonaphthalene-2-carboxylate

  • MDL number MFCD12159841
  • Cas Number : 35092-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95098
Availability details for
Quantity
  • Availability
  • Add to Cart