ENA369383389 Enamine

2-bromo-6-[(methylamino)methyl]phenol

  • MDL number MFCD14656483
  • Cas Number : 157729-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102496

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102496
Availability details for
Quantity
  • Availability
  • Add to Cart