ENA369383395 Enamine

4-(chloromethyl)-2-[1-(2-methylpropoxy)ethyl]-1,3-thiazole

  • MDL number MFCD16079182
  • Cas Number : 1247506-33-7
  • Molecular Weight : 233.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96406
Availability details for
Quantity
  • Availability
  • Add to Cart