ENA369383400 Enamine

2,2,3,3-tetrafluoropropanoic acid

  • MDL number MFCD00054686
  • Cas Number : 756-09-2
  • Molecular Weight : 146.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96639

Safety Information

signalword Danger
pcodes P234,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P390,P403+P233,P405,P406,P501
MSDS https://www.enaminestore.com/catalog/EN300-96639
pictograms GHS05,GHS07
hstatements H290,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart