ENA369383402 Enamine

2-(5-chloro-2-methoxyphenyl)-2,2-difluoroacetic acid

  • MDL number MFCD16748870
  • Cas Number : 1249203-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102497
Availability details for
Quantity
  • Availability
  • Add to Cart