ENA369383404 Enamine

1-(aminomethyl)-N-(3,4-dimethylphenyl)cyclopentane-1-carboxamide

  • MDL number MFCD16541442
  • Cas Number : 1305480-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-96812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96812
Availability details for
Quantity
  • Availability
  • Add to Cart