ENA369383428 Enamine

2,4-difluoro-5-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD22196689
  • Cas Number : 1235407-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97511

Safety Information

signalword Danger
pcodes P234,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P390,P405,P406,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-97511
pictograms GHS05
hstatements H290,H314
Availability details for
Quantity
  • Availability
  • Add to Cart