ENA369383429 Enamine

{1-[(2-phenylethyl)amino]cyclopentyl}methanol

  • MDL number MFCD16784451
  • Cas Number : 1250923-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102872
Availability details for
Quantity
  • Availability
  • Add to Cart