ENA369383435 Enamine

{1-[(pyrrolidin-2-yl)methyl]pyrrolidin-2-yl}methanol

  • MDL number MFCD22056472
  • Cas Number : 1375987-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97712
Availability details for
Quantity
  • Availability
  • Add to Cart