ENA369383451 Enamine

4-(methylsulfamoyl)thiophene-2-carboxylic acid

  • MDL number MFCD03838937
  • Cas Number : 667901-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98100
Availability details for
Quantity
  • Availability
  • Add to Cart