ENA369383463 Enamine

1,3-thiazole-5-carbonitrile

  • MDL number MFCD06797206
  • Cas Number : 25742-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98533

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98533
Availability details for
Quantity
  • Availability
  • Add to Cart