ENA369383473 Enamine

5-chloro-1-(3-chloro-4-methylphenyl)-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11540962
  • Cas Number : 1152574-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241...243
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102928

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102928
Availability details for
Quantity
  • Availability
  • Add to Cart