ENA369383479 Enamine

2,4,4-trimethylcyclohexan-1-amine

  • MDL number MFCD22056477
  • Cas Number : 1375982-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98940
Availability details for
Quantity
  • Availability
  • Add to Cart