ENA369383481 Enamine

3-methyl-2-(pyridin-3-yl)butan-1-amine

  • MDL number MFCD21909408
  • Cas Number : 1368841-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98977
Availability details for
Quantity
  • Availability
  • Add to Cart