ENA369383493 Enamine

5-methyl-5-(pyridin-3-yl)imidazolidine-2,4-dione

  • MDL number MFCD09741362
  • Cas Number : 54743-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99236
Availability details for
Quantity
  • Availability
  • Add to Cart