ENA369383497 Enamine

4-(4H-1,2,4-triazol-3-yl)piperidine

  • MDL number MFCD14581791
  • Cas Number : 893424-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99247

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-99247
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart