ENA369383505 Enamine

1-(3-aminophenyl)-4-methylpentan-3-one

  • MDL number MFCD22196694
  • Cas Number : 1375989-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99353
Availability details for
Quantity
  • Availability
  • Add to Cart