ENA369383517 Enamine

methyl 2-{[(5-chlorothiophen-2-yl)methyl]amino}acetate

  • MDL number MFCD11150560
  • Cas Number : 1021000-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99447
Availability details for
Quantity
  • Availability
  • Add to Cart