ENA369383528 Enamine

2-(bromomethyl)bicyclo[2.2.1]heptane

  • MDL number MFCD16860589
  • Cas Number : 55932-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99571

Safety Information

signalword Warning
MSDS https://www.enaminestore.com/catalog/EN300-99571
pictograms GHS07
hstatements EUH209/209A,H227,H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart