ENA369383529 Enamine

1-methoxy-2-phenylpropan-2-ol

  • MDL number MFCD16787658
  • Cas Number : 3587-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99573
Availability details for
Quantity
  • Availability
  • Add to Cart