ENA369383530 Enamine

2-{[4-amino-5-(5-methyl-1-benzofuran-2-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD22056486
  • Cas Number : 1304271-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99580
Availability details for
Quantity
  • Availability
  • Add to Cart