ENA369383532 Enamine

tert-butyl 3-hydroxy-3-methylazetidine-1-carboxylate

  • MDL number MFCD16037112
  • Cas Number : 1104083-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99594
Availability details for
Quantity
  • Availability
  • Add to Cart