ENA369383534 Enamine

3-(ethylsulfanyl)cyclohexan-1-amine

  • MDL number MFCD20402579
  • Cas Number : 1340433-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99603
Availability details for
Quantity
  • Availability
  • Add to Cart