ENA369383541 Enamine

3-ethoxy-2,2-diethylcyclobutan-1-ol

  • MDL number MFCD21647897
  • Cas Number : 1376341-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99636
Availability details for
Quantity
  • Availability
  • Add to Cart