ENA369383546 Enamine

(3-methoxy-2-methylpropyl)(methyl)amine

  • MDL number MFCD16148617
  • Cas Number : 1247523-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99641
Availability details for
Quantity
  • Availability
  • Add to Cart