ENA369383553 Enamine

2-(dimethylamino)-2-(1H-pyrrol-3-yl)acetonitrile

  • MDL number MFCD12964755
  • Cas Number : 117067-99-9
  • Molecular Weight : 149.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-99676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99676
Availability details for
Quantity
  • Availability
  • Add to Cart