ENA369383559 Enamine

5,7-dichloro-1,3-benzoxazol-2-amine

  • MDL number MFCD16742784
  • Cas Number : 98555-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247...249
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99682

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99682
Availability details for
Quantity
  • Availability
  • Add to Cart