ENA369383560 Enamine

(1-benzoylpiperidin-3-yl)methanol

  • MDL number MFCD00783787
  • Cas Number : 145021-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-99683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99683
Availability details for
Quantity
  • Availability
  • Add to Cart