ENA369383562 Enamine

1-(3-methoxyphenyl)-2,2-dimethylpropan-1-amine

  • MDL number MFCD12154104
  • Cas Number : 654683-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99685

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99685
Availability details for
Quantity
  • Availability
  • Add to Cart