ENA369383569 Enamine

5-(3-chloro-4-fluorophenyl)-1H-pyrazole

  • MDL number MFCD03407968
  • Cas Number : 154258-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99763

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99763
Availability details for
Quantity
  • Availability
  • Add to Cart