ENA369383570 Enamine

4-[(oxan-2-yl)methoxy]pyridine

  • MDL number MFCD22056498
  • Cas Number : 1376224-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99765
Availability details for
Quantity
  • Availability
  • Add to Cart