ENA369383573 Enamine

2-(1-benzofuran-2-yl)propanoic acid

  • MDL number MFCD14602275
  • Cas Number : 1249571-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-60
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99769
Availability details for
Quantity
  • Availability
  • Add to Cart