ENA369383575 Enamine

N-(3-aminopropyl)cyclobutanecarboxamide hydrochloride

  • MDL number MFCD22056500
  • Cas Number : 1243268-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99774
Availability details for
Quantity
  • Availability
  • Add to Cart