ENA369383579 Enamine

2-(N'-hydroxycarbamimidoyl)-N-methylacetamide

  • Cas Number : 1094362-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99778
Availability details for
Quantity
  • Availability
  • Add to Cart