ENA369383580 Enamine

ethyl 2-(3-carbamothioylphenoxy)acetate

  • MDL number MFCD09949046
  • Cas Number : 1016841-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99779

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99779
Availability details for
Quantity
  • Availability
  • Add to Cart