ENA369383581 Enamine

1-[(3-chlorophenyl)methyl]-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD18556599
  • Cas Number : 1376361-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-99781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99781
Availability details for
Quantity
  • Availability
  • Add to Cart