ENA369383582 Enamine

ethyl 2-(4-carbamothioylphenoxy)acetate

  • MDL number MFCD09936031
  • Cas Number : 1017036-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99785
Availability details for
Quantity
  • Availability
  • Add to Cart