ENA369383586 Enamine

1-(3-bromopropyl)-4-(trifluoromethyl)benzene

  • MDL number MFCD09744406
  • Cas Number : 178369-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99803
Availability details for
Quantity
  • Availability
  • Add to Cart