ENA369383587 Enamine

2-(1,2,3,4-tetrahydronaphthalen-1-yl)pyrrolidine

  • MDL number MFCD16857962
  • Cas Number : 95092-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99804

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99804
Availability details for
Quantity
  • Availability
  • Add to Cart