ENA369383588 Enamine

(2,2,3,3-tetramethylcyclopropyl)methanol

  • MDL number MFCD21783553
  • Cas Number : 2415-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99805
Availability details for
Quantity
  • Availability
  • Add to Cart