ENA369383607 Enamine

2-{N-methyl-1-[1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazol-4-yl]formamido}acetic acid

  • MDL number MFCD12571019
  • Cas Number : 1152952-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99817
Availability details for
Quantity
  • Availability
  • Add to Cart