ENA369383609 Enamine

4-[(furan-2-yl)carbonyl]-3-methylpiperazin-2-one

  • MDL number MFCD09987262
  • Cas Number : 1103994-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99822

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99822
Availability details for
Quantity
  • Availability
  • Add to Cart