ENA369383623 Enamine

2-(3-nitro-1H-1,2,4-triazol-1-yl)ethan-1-ol

  • MDL number MFCD03059897
  • Cas Number : 105958-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99878
Availability details for
Quantity
  • Availability
  • Add to Cart