ENA369383633 Enamine

(cyclopropylmethyl)(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD22056511
  • Cas Number : 405878-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99895
Availability details for
Quantity
  • Availability
  • Add to Cart