ENA369383634 Enamine

2-(4-fluorophenyl)piperidine hydrochloride

  • MDL number MFCD02178957
  • Cas Number : 1187174-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99896
Availability details for
Quantity
  • Availability
  • Add to Cart