ENA369383636 Enamine

6-(4-aminophenyl)-2,3,4,5-tetrahydropyridazin-3-one

  • MDL number MFCD00266152
  • Cas Number : 21282-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99898
Availability details for
Quantity
  • Availability
  • Add to Cart