ENA369383637 Enamine

2-(pyridazin-3-ylamino)ethan-1-ol

  • MDL number MFCD16767602
  • Cas Number : 1247645-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99899

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99899
Availability details for
Quantity
  • Availability
  • Add to Cart