ENA369383638 Enamine

(1-propoxycyclohexyl)methanamine hydrochloride

  • MDL number MFCD22056512
  • Cas Number : 1376352-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99900
Availability details for
Quantity
  • Availability
  • Add to Cart