ENA369383649 Enamine

1-[(1-aminobutan-2-yl)oxy]-2-chlorobenzene

  • MDL number MFCD06246391
  • Cas Number : 883519-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99920

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99920
Availability details for
Quantity
  • Availability
  • Add to Cart