ENA369383652 Enamine

(3,4-dihydro-1H-2-benzothiopyran-1-yl)methanamine

  • MDL number MFCD20698414
  • Cas Number : 19158-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99923
Availability details for
Quantity
  • Availability
  • Add to Cart